Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009885.1 | NP_034015.1 | 8 - 9 | 1087 | 59 |
GenBank mRNA | AK131818.1 | - | 8 - 9 | 1105 | 59 |
AK131824.1 | - | 8 - 9 | 1105 | 59 | |
BC006872.1 | - | 8 - 9 | 1099 | 59 | |
U33169.1 | - | 8 - 9 | 1087 | 59 | |
U37386.1 | - | 8 - 9 | 1091 | 59 |