Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177352.1 | NP_001170823.1 | 1 - 2 | 611 | 83 |
NM_010849.4 | NP_034979.3 | 1 - 2 | 611 | 83 | |
GenBank mRNA | AK087961.1 | - | 1 - 2 | 403 | 83 |
AK145084.1 | - | 1 - 2 | 565 | 83 | |
BC138931.1 | - | 1 - 2 | 74 | 83 | |
BC138932.1 | - | 1 - 2 | 74 | 83 | |
X01023.1 | - | 1 - 2 | 591 | 83 |