Assay Design



Gene Transcripts

Gene Symbol
Rad51d
Entrez Gene ID
19364
Gene Name
RAD51 homolog D
Gene Aliases
R51H3, Rad51l3, TRAD
Chromosome Location
Chr.11: 82871960 - 82890751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277938.1 NP_001264867.1 4 - 5 546 99
NM_001277939.1 NP_001264868.1 4 - 5 546 99
NM_001277942.1 NP_001264871.1 4 - 5 546 99
NM_011235.4 NP_035365.1 4 - 5 546 99
NR_102717.1 4 - 5 546 99
NR_102718.1 4 - 5 569 99
NR_102719.1 3 - 4 427 99
NR_102720.1 3 - 4 427 99
GenBank mRNA AB007040.1 - 4 - 5 511 99
AB052828.1 - 4 - 5 511 99
AB052829.1 - 4 - 5 511 99
AB052830.1 - 4 - 5 511 99
AB052831.1 - 3 - 4 392 99
AB052832.1 - 3 - 4 392 99
AB052834.1 - 4 - 5 534 99
AF034955.1 - 4 - 5 536 99
AK017835.1 - 4 - 5 529 99
AK036272.1 - 3 - 4 385 99
AK138553.1 - 4 - 5 538 99
AK147759.1 - 4 - 5 521 99
BC049136.1 - 4 - 5 511 99
EU627687.1 - 4 - 5 536 99
EU627688.1 - 4 - 5 536 99
Y15570.1 - 4 - 5 532 99

View Full Product Details