Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277938.1 | NP_001264867.1 | 9 - 10 | 1096 | 73 |
NM_001277941.1 | NP_001264870.1 | 8 - 9 | 976 | 73 | |
NM_001277942.1 | NP_001264871.1 | 7 - 8 | 949 | 73 | |
NM_011235.4 | NP_035365.1 | 9 - 10 | 1111 | 73 | |
NR_102717.1 | 8 - 9 | 1040 | 73 | ||
NR_102718.1 | 9 - 10 | 1134 | 73 | ||
NR_102719.1 | 8 - 9 | 992 | 73 | ||
NR_102720.1 | 8 - 9 | 977 | 73 | ||
GenBank mRNA | AB007040.1 | - | 9 - 10 | 1076 | 73 |
AB052828.1 | - | 8 - 9 | 1005 | 73 | |
AB052829.1 | - | 9 - 10 | 1061 | 73 | |
AB052830.1 | - | 7 - 8 | 914 | 73 | |
AB052831.1 | - | 8 - 9 | 957 | 73 | |
AB052832.1 | - | 8 - 9 | 942 | 73 | |
AB052833.1 | - | 8 - 9 | 941 | 73 | |
AB052834.1 | - | 9 - 10 | 1099 | 73 | |
AF034955.1 | - | 9 - 10 | 1101 | 73 | |
AK017835.1 | - | 9 - 10 | 1094 | 73 | |
AK036272.1 | - | 8 - 9 | 950 | 73 | |
AK138553.1 | - | 9 - 10 | 1103 | 73 | |
AK147759.1 | - | 9 - 10 | 1086 | 73 | |
BC049136.1 | - | 9 - 10 | 1076 | 73 | |
CF746344.1 | - | 5 - 6 | 528 | 73 | |
Y15570.1 | - | 9 - 10 | 1097 | 73 |