Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011237.2 | NP_035367.1 | 5 - 6 | 539 | 90 |
GenBank mRNA | AF045663.1 | - | 5 - 6 | 539 | 90 |
AK134293.1 | - | 5 - 6 | 552 | 90 | |
AK138973.1 | - | 5 - 6 | 544 | 90 | |
AK165833.1 | - | 5 - 6 | 574 | 90 | |
AK186361.1 | - | 3 - 4 | 265 | 90 | |
AK219932.1 | - | 1 - 2 | 50 | 90 | |
BC082556.1 | - | 5 - 6 | 478 | 90 |