Assay Design



Gene Transcripts

Gene Symbol
Sbp
Entrez Gene ID
20234
Gene Name
spermine binding protein
Gene Aliases
AV082137, AV082259, p25
Chromosome Location
Chr.17: 23941996 - 23945607 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011321.3 NP_035451.1 2 - 3 238 86
GenBank mRNA AK035423.1 - 2 - 3 239 86
BC049605.1 - 2 - 3 238 86
X06246.1 - 2 - 3 238 86

View Full Product Details