Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011355.1 | NP_035485.1 | 1 - 2 | 194 | 82 |
GenBank mRNA | AK142246.1 | - | 1 - 2 | 211 | 82 |
AK153524.1 | - | 1 - 2 | 225 | 82 | |
AK154945.1 | - | 1 - 2 | 246 | 82 | |
AK155085.1 | - | 1 - 2 | 230 | 82 | |
BC003815.1 | - | 1 - 2 | 112 | 82 | |
L03215.1 | - | 1 - 2 | 91 | 82 | |
M32370.1 | - | 1 - 2 | 174 | 82 | |
M38252.1 | - | 1 - 2 | 194 | 82 | |
X17463.1 | - | 1 - 2 | 882 | 82 |