Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011422.3 | NP_035552.1 | 2 - 3 | 153 | 64 |
GenBank mRNA | AV481993.1 | - | - | 244 | 64 |
BC031806.1 | - | 2 - 3 | 119 | 64 | |
BC031921.1 | - | 2 - 3 | 97 | 64 | |
BC034553.1 | - | 2 - 3 | 123 | 64 | |
BC035428.1 | - | 2 - 3 | 125 | 64 | |
BC055857.1 | - | 2 - 3 | 147 | 64 | |
BC141243.1 | - | 2 - 3 | 115 | 64 | |
BC145569.1 | - | 2 - 3 | 115 | 64 | |
BF454619.1 | - | 2 - 3 | 121 | 64 | |
X71629.1 | - | 2 - 3 | 130 | 64 | |
X71631.1 | - | - | 130 | 64 |