Assay Design



Gene Transcripts

Gene Symbol
Figla
Entrez Gene ID
26910
Gene Name
folliculogenesis specific basic helix-loop-helix
Gene Aliases
bHLHc8
Chromosome Location
Chr.6: 86010320 - 86020996 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012013.1 NP_036143.1 2 - 3 422 106
GenBank mRNA JN700518.1 - 2 - 3 382 106
U91840.1 - 2 - 3 422 106

View Full Product Details