Assay Design
Gene Transcripts
- Gene Symbol
- Sh3d19
- Entrez Gene ID
-
27059
- Gene Name
- SH3 domain protein D19
- Gene Aliases
-
AW011754, Kryn
- Chromosome Location
-
Chr.3: 85970956 - 86130522 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D89677.2 |
- |
3 - 4 |
103 |
70 |
View Full Product Details