Assay Design



Gene Transcripts

Gene Symbol
Cbx8
Entrez Gene ID
30951
Gene Name
chromobox 8
Gene Aliases
Pc3
Chromosome Location
Chr.11: 119038436 - 119040913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013926.1 NP_038954.1 1 - 2 133 58
GenBank mRNA AB221673.1 - 1 - 2 70 58
AF180370.1 - 1 - 2 133 58
BC014815.1 - 1 - 2 83 58

View Full Product Details