Assay Design
Gene Transcripts
- Gene Symbol
- Cngb3
- Entrez Gene ID
-
30952
- Gene Name
- cyclic nucleotide gated channel beta 3
- Gene Aliases
-
CCNC2, CNG6, Cngbeta2
- Chromosome Location
-
Chr.4: 19280850 - 19510623 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_013927.2 |
NP_038955.1 |
8 - 9 |
1041 |
107 |
GenBank mRNA |
AJ243572.1 |
- |
8 - 9 |
1041 |
107 |
View Full Product Details