Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016911.4 | NP_058607.1 | 8 - 9 | 1215 | 61 |
GenBank mRNA | AB028049.1 | - | 8 - 9 | 1142 | 61 |
AB028050.1 | - | 6 - 7 | 890 | 61 | |
AK034696.1 | - | 8 - 9 | 1217 | 61 | |
AK159662.1 | - | 7 - 8 | 1203 | 61 | |
BC022572.1 | - | 8 - 9 | 1171 | 61 |