Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166531.1 | NP_001160003.1 | 20 - 21 | 2599 | 114 |
NM_001166532.1 | NP_001160004.1 | 19 - 20 | 2693 | 114 | |
NM_019460.2 | NP_062333.1 | 19 - 20 | 2550 | 114 | |
GenBank mRNA | AB032165.1 | - | 18 - 19 | 2395 | 114 |