Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168505.1 | NP_001161977.1 | 7 - 8 | 1746 | 65 |
NM_019658.6 | NP_062632.2 | 7 - 8 | 1787 | 65 | |
GenBank mRNA | AF068921.1 | - | 6 - 7 | 1422 | 65 |
AK146447.1 | - | 7 - 8 | 1784 | 65 | |
BC013722.1 | - | 3 - 4 | 322 | 65 | |
BC026364.1 | - | 6 - 7 | 1653 | 65 | |
BC049775.1 | - | 6 - 7 | 1652 | 65 | |
BC083060.1 | - | 7 - 8 | 1746 | 65 |