Assay Design



Gene Transcripts

Gene Symbol
Mbtps1
Entrez Gene ID
56453
Gene Name
membrane-bound transcription factor peptidase, site 1
Gene Aliases
0610038M03Rik, AV003995, S1P, SKI-1, Ski1, mKIAA0091
Chromosome Location
Chr.8: 119508151 - 119558825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167910.1 NP_001161382.1 18 - 19 2867 76
NM_019709.4 NP_062683.3 19 - 20 3016 76
GenBank mRNA AF094820.1 - 17 - 18 2435 76
AK029048.1 - 18 - 19 2844 76
AK122202.1 - 18 - 19 2833 76
AK147336.1 - 17 - 18 5436 76
AK147495.1 - 18 - 19 2841 76
BC011533.1 - 18 - 19 2830 76
BC054837.1 - 17 - 18 2635 76
BC057198.1 - 19 - 20 2982 76
DQ913734.1 - 17 - 18 2498 76

View Full Product Details