Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019775.3 | NP_062749.2 | 1 - 2 | 137 | 83 |
GenBank mRNA | AA048136.1 | - | 1 - 2 | 137 | 83 |
AB021968.1 | - | 1 - 2 | 85 | 83 | |
AF164524.1 | - | 1 - 2 | 152 | 83 | |
AF186188.1 | - | 1 - 2 | 88 | 83 | |
AK004045.1 | - | 1 - 2 | 110 | 83 | |
BC089577.1 | - | 1 - 2 | 109 | 83 |