Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019775.3 | NP_062749.2 | 9 - 10 | 1061 | 141 |
GenBank mRNA | AB021968.1 | - | 9 - 10 | 1009 | 141 |
AF164524.1 | - | 9 - 10 | 1076 | 141 | |
AF186188.1 | - | 9 - 10 | 1012 | 141 | |
AK004045.1 | - | 9 - 10 | 1034 | 141 | |
BC089577.1 | - | 9 - 10 | 1033 | 141 |