Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020593.2 | NP_065618.1 | 1 - 2 | 161 | 82 |
NM_212433.1 | NP_997598.1 | 1 - 2 | 161 | 82 | |
GenBank mRNA | AF233226.1 | - | 1 - 2 | 100 | 82 |
AK004544.2 | - | 1 - 2 | 113 | 82 | |
AK019550.1 | - | 1 - 2 | 101 | 82 | |
AK028048.1 | - | 1 - 2 | 123 | 82 | |
AK031185.1 | - | 1 - 2 | 100 | 82 | |
AK031428.1 | - | 1 - 2 | 121 | 82 | |
AK049870.1 | - | 1 - 2 | 119 | 82 | |
AK050195.1 | - | 1 - 2 | 113 | 82 | |
AK085686.1 | - | 1 - 2 | 116 | 82 | |
AK150097.1 | - | 1 - 2 | 113 | 82 | |
AK151130.1 | - | 1 - 2 | 112 | 82 | |
AK151857.1 | - | 1 - 2 | 112 | 82 | |
AK152780.1 | - | 1 - 2 | 112 | 82 | |
AK167075.1 | - | 1 - 2 | 99 | 82 | |
AK215292.1 | - | 1 - 2 | 105 | 82 | |
AK215501.1 | - | 1 - 2 | 105 | 82 | |
BC010212.1 | - | 1 - 2 | 62 | 82 | |
BC050758.1 | - | 1 - 2 | 114 | 82 | |
BC058253.1 | - | 1 - 2 | 118 | 82 |