Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025633.3 | NP_079909.1 | 2 - 3 | 222 | 137 |
GenBank mRNA | AK005241.1 | - | 2 - 3 | 219 | 137 |
AK010067.1 | - | 2 - 3 | 222 | 137 | |
BC060739.1 | - | 2 - 3 | 209 | 137 |