Assay Design



Gene Transcripts

Gene Symbol
Cd82
Entrez Gene ID
12521
Gene Name
CD82 antigen
Gene Aliases
AA682076, AL023070, C33, IA4, Kai1, Tspan27
Chromosome Location
Chr.2: 93419102 - 93462950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271430.1 NP_001258359.1 1 - 2 175 108
GenBank mRNA AK160907.1 - 1 - 2 97 108
D14883.1 - 1 - 2 84 108

View Full Product Details