Assay Design



Gene Transcripts

Gene Symbol
Lamb2
Entrez Gene ID
16779
Gene Name
laminin, beta 2
Gene Aliases
AW211941, Lamb-2, Lams, npht
Chromosome Location
Chr.9: 108479857 - 108490530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008483.3 NP_032509.2 6 - 7 770 101
GenBank mRNA AK140351.1 - 6 - 7 749 101
AK158444.1 - 6 - 7 742 101
AK193413.1 - 1 - 2 35 101
BC026051.1 - 6 - 7 697 101

View Full Product Details