Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166584.1 | NP_001160056.1 | 7 - 8 | 1003 | 68 |
NM_001166585.1 | NP_001160057.1 | 4 - 5 | 521 | 68 | |
NM_009346.3 | NP_033372.1 | 6 - 7 | 940 | 68 | |
GenBank mRNA | AK140307.1 | - | 7 - 8 | 1004 | 68 |
AK148375.1 | - | 6 - 7 | 906 | 68 | |
BC052021.1 | - | 4 - 5 | 462 | 68 | |
BC060138.1 | - | 4 - 5 | 462 | 68 | |
L06865.1 | - | 4 - 5 | 467 | 68 | |
L13853.1 | - | 6 - 7 | 667 | 68 | |
S74227.1 | - | 5 - 6 | 928 | 68 |