Assay Design



Gene Transcripts

Gene Symbol
Adad1
Entrez Gene ID
21744
Gene Name
adenosine deaminase domain containing 1 (testis specific)
Gene Aliases
Tenr
Chromosome Location
Chr.3: 37063093 - 37121930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009350.3 NP_033376.2 8 - 9 1215 68
GenBank mRNA AK133008.1 - 5 - 6 631 68
X84693.1 - 8 - 9 1203 68

View Full Product Details