Assay Design



Gene Transcripts

Gene Symbol
Top1
Entrez Gene ID
21969
Gene Name
topoisomerase (DNA) I
Gene Aliases
AI467334, D130064I21Rik, Top-1
Chromosome Location
Chr.2: 160645897 - 160722764 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009408.2 NP_033434.2 17 - 18 2164 86
GenBank mRNA D10061.1 - 17 - 18 1831 86
L20632.1 - 17 - 18 1828 86

View Full Product Details