Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009421.3 | NP_033447.2 | 9 - 10 | 1338 | 63 |
GenBank mRNA | AK029135.1 | - | 9 - 10 | 1340 | 63 |
AK145075.1 | - | - | 913 | 63 | |
AK169515.1 | - | 9 - 10 | 1131 | 63 | |
AK172598.1 | - | 5 - 6 | 942 | 63 | |
BC096595.1 | - | 5 - 6 | 904 | 63 | |
L35302.1 | - | 8 - 9 | 1084 | 63 |