Assay Design



Gene Transcripts

Gene Symbol
Vpreb3
Entrez Gene ID
22364
Gene Name
pre-B lymphocyte gene 3
Gene Aliases
8HS-20, AI528709, Vpreb-3
Chromosome Location
Chr.10: 75943025 - 75949651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009514.4 NP_033540.1 1 - 2 95 63
GenBank mRNA AK008794.1 - 1 - 2 96 63
BC062250.1 - 1 - 2 75 63

View Full Product Details