Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010023.4 | NP_034153.2 | 3 - 4 | 281 | 78 |
GenBank mRNA | AK004838.1 | - | 3 - 4 | 282 | 78 |
AK029481.1 | - | 3 - 4 | 282 | 78 | |
AK077386.1 | - | 3 - 4 | 272 | 78 | |
BC022712.1 | - | 3 - 4 | 265 | 78 | |
BC054444.1 | - | 3 - 4 | 268 | 78 | |
Z14049.1 | - | 3 - 4 | 261 | 78 |