Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278226.1 | NP_001265155.1 | 10 - 11 | 849 | 80 |
NM_010059.3 | NP_034189.1 | 12 - 13 | 1014 | 80 | |
NR_103477.1 | 7 - 8 | 639 | 80 | ||
GenBank mRNA | AK076777.1 | - | 10 - 11 | 840 | 80 |
AK169570.1 | - | 7 - 8 | 639 | 80 | |
BC116767.1 | - | 12 - 13 | 899 | 80 | |
BC119081.1 | - | 12 - 13 | 899 | 80 | |
D58419.1 | - | 12 - 13 | 999 | 80 | |
D64107.1 | - | 12 - 13 | 988 | 80 |