Assay Design



Gene Transcripts

Gene Symbol
Cd47
Entrez Gene ID
16423
Gene Name
CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
Gene Aliases
9130415E20Rik, AA407862, AI848868, AW108519, B430305P08Rik, IAP, Itgp
Chromosome Location
Chr.16: 49855252 - 49912424 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010581.3 NP_034711.1 3 - 4 588 102
GenBank mRNA AB012693.1 - 3 - 4 527 102
BC012667.1 - 3 - 4 522 102
Z25524.1 - 3 - 4 588 102

View Full Product Details