Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010684.2 | NP_034814.2 | 4 - 5 | 799 | 76 |
GenBank mRNA | AK004637.1 | - | 4 - 5 | 666 | 76 |
AK159096.1 | - | 4 - 5 | 665 | 76 | |
AK159376.1 | - | 4 - 5 | 638 | 76 | |
AK172007.1 | - | 4 - 5 | 681 | 76 | |
AK181514.1 | - | 1 - 2 | 53 | 76 | |
AK183713.1 | - | 1 - 2 | 53 | 76 | |
AK184936.1 | - | 1 - 2 | 86 | 76 | |
AK186339.1 | - | 1 - 2 | 53 | 76 | |
AK200388.1 | - | 1 - 2 | 53 | 76 | |
AK206801.1 | - | 1 - 2 | 159 | 76 | |
AK210006.1 | - | 1 - 2 | 77 | ||
AK215533.1 | - | 1 - 2 | 53 | 76 | |
AY069968.1 | - | 4 - 5 | 736 | 76 | |
BC049097.1 | - | 4 - 5 | 773 | 76 | |
J03881.1 | - | 3 - 4 | 473 | 76 | |
M25244.1 | - | 4 - 5 | 654 | 76 | |
M32015.1 | - | 4 - 5 | 623 | 76 |