Assay Design



Gene Transcripts

Gene Symbol
Lig1
Entrez Gene ID
16881
Gene Name
ligase I, DNA, ATP-dependent
Gene Aliases
AL033288, LigI
Chromosome Location
Chr.7: 13277313 - 13311427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083188.1 NP_001076657.1 16 - 17 1787 78
NM_001199310.1 NP_001186239.1 16 - 17 1795 78
NM_010715.2 NP_034845.2 16 - 17 1712 78
GenBank mRNA AK153993.1 - 16 - 17 1674 78
AK160208.1 - 1 - 2 81 78
AK161793.1 - 9 - 10 870 78
AK168643.1 - 16 - 17 1790 78
BC028287.1 - 16 - 17 1637 78
BC138541.1 - 16 - 17 1795 78
BC138542.1 - 16 - 17 1795 78
U04674.1 - 15 - 16 1580 78
U19604.1 - 16 - 17 1825 78

View Full Product Details