Assay Design



Gene Transcripts

Gene Symbol
Lig1
Entrez Gene ID
16881
Gene Name
ligase I, DNA, ATP-dependent
Gene Aliases
AL033288, LigI
Chromosome Location
Chr.7: 13277313 - 13311427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083188.1 NP_001076657.1 23 - 24 2493 61
NM_001199310.1 NP_001186239.1 23 - 24 2501 61
NM_010715.2 NP_034845.2 23 - 24 2418 61
GenBank mRNA AK153993.1 - 23 - 24 2380 61
AK160208.1 - 8 - 9 787 61
AK161793.1 - 16 - 17 1576 61
AK168643.1 - 23 - 24 2497 61
BC028287.1 - 23 - 24 2343 61
BC138541.1 - 23 - 24 2501 61
BC138542.1 - 23 - 24 2501 61
U04674.1 - 22 - 23 2286 61
U19604.1 - 23 - 24 2531 61

View Full Product Details