Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011818.3 | NP_035948.3 | 3 - 4 | 690 | 73 |
GenBank mRNA | AF163665.1 | - | 3 - 4 | 634 | 73 |
AF186095.1 | - | 3 - 4 | 526 | 73 | |
AK004716.1 | - | 3 - 4 | 666 | 73 | |
AK132788.1 | - | 3 - 4 | 554 | 73 | |
AK146964.1 | - | 3 - 4 | 659 | 73 | |
AK155238.1 | - | 3 - 4 | 673 | 73 | |
BC054767.1 | - | 3 - 4 | 649 | 73 |