Assay Design



Gene Transcripts

Gene Symbol
Aadat
Entrez Gene ID
23923
Gene Name
aminoadipate aminotransferase
Gene Aliases
AI875679, Aadt, KATII, Kat2, mKat-2
Chromosome Location
Chr.8: 60505870 - 60545677 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011834.2 NP_035964.1 1 - 2 159 72
GenBank mRNA AF072376.1 - 1 - 2 159 72
AK075578.1 - 1 - 2 165 72
BC012637.1 - 1 - 2 162 72

View Full Product Details