Assay Design



Gene Transcripts

Gene Symbol
Ankrd1
Entrez Gene ID
107765
Gene Name
ankyrin repeat domain 1 (cardiac muscle)
Gene Aliases
Alrp, CARP, Crap, MARP1
Chromosome Location
Chr.19: 36111965 - 36119844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013468.3 NP_038496.2 6 - 7 718 70
GenBank mRNA AF041847.1 - 6 - 7 669 70
AK009655.1 - 6 - 7 711 70
AK009959.1 - 6 - 7 717 70
AK145944.1 - 6 - 7 759 70
AK146471.1 - 6 - 7 715 70
AK146627.1 - 6 - 7 717 70
AK146940.1 - 6 - 7 715 70
BC037138.1 - 6 - 7 718 70

View Full Product Details