Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013705.1 | NP_038733.1 | 3 - 4 | 323 | 71 |
GenBank mRNA | AK014064.1 | - | 2 - 3 | 165 | 71 |
AY605122.1 | - | 3 - 4 | 323 | 71 | |
BC049120.1 | - | 3 - 4 | 324 | 71 | |
BC051972.1 | - | 3 - 4 | 523 | 71 | |
Z30174.1 | - | 3 - 4 | 323 | 71 |