Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110305.1 | NP_001103775.1 | 2 - 3 | 1232 | 71 |
NM_001110306.1 | NP_001103776.1 | 2 - 3 | 1117 | 71 | |
NM_001110307.1 | NP_001103777.1 | 2 - 3 | 949 | 71 | |
NM_016679.4 | NP_057888.1 | 1 - 2 | 2268 | 71 | |
GenBank mRNA | AB020063.1 | - | 2 - 3 | 721 | 71 |
AK004738.1 | - | 2 - 3 | 949 | 71 | |
AK039549.1 | - | 2 - 3 | 888 | 71 | |
AK046178.1 | - | 2 - 3 | 833 | 71 | |
AK076234.1 | - | 1 - 2 | 1754 | 71 | |
AK129061.1 | - | 1 - 2 | 703 | 71 | |
AK150437.1 | - | 2 - 3 | 853 | 71 | |
AK150485.1 | - | 2 - 3 | 720 | 71 | |
AK152142.1 | - | 2 - 3 | 853 | 71 | |
AK154430.1 | - | 2 - 3 | 764 | 71 | |
AK155507.1 | - | 2 - 3 | 764 | 71 | |
AK159858.1 | - | 2 - 3 | 718 | 71 | |
BC006729.1 | - | 2 - 3 | 730 | 71 | |
BC023945.1 | - | 2 - 3 | 982 | 71 | |
BC055732.1 | - | 1 - 2 | 706 | 71 |