Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018824.2 | NP_061294.2 | 2 - 3 | 349 | 56 |
GenBank mRNA | AB038145.1 | - | 1 - 2 | 85 | 56 |
AK087175.1 | - | 2 - 3 | 357 | 56 | |
AK169991.1 | - | 2 - 3 | 351 | 56 | |
AY004874.1 | - | 2 - 3 | 303 | 56 | |
BC050823.1 | - | 1 - 2 | 113 | 56 |