Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020046.3 | NP_064430.1 | 8 - 9 | 1432 | 70 |
GenBank mRNA | AF029667.2 | - | 8 - 9 | 1137 | 70 |
AK030798.1 | - | - | 1016 | 70 | |
BC019542.1 | - | 8 - 9 | 1142 | 70 | |
BC027829.1 | - | 8 - 9 | 1393 | 70 | |
BC045206.1 | - | 8 - 9 | 1382 | 70 |