Assay Design
Gene Transcripts
- Gene Symbol
- Dnajc4
- Entrez Gene ID
-
57431
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 4
- Gene Aliases
-
2010301J22Rik, Hspf2, Mcg18
- Chromosome Location
-
Chr.19: 6987911 - 6992696 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_020566.1 |
NP_065591.1 |
3 - 4 |
390 |
90 |
GenBank mRNA |
AF036875.1 |
- |
3 - 4 |
390 |
90 |
View Full Product Details