Assay Design



Gene Transcripts

Gene Symbol
Wdr4
Entrez Gene ID
57773
Gene Name
WD repeat domain 4
Gene Aliases
AI415180, AI448349, D530049K22Rik
Chromosome Location
Chr.17: 31494322 - 31519992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021322.2 NP_067297.2 2 - 3 391 64
GenBank mRNA AJ271892.1 - 2 - 3 161 64
AJ271893.1 - 2 - 3 161 64
AK035125.1 - 2 - 3 172 64
AK144821.1 - 2 - 3 196 64
AK152271.1 - 2 - 3 183 64
AK153161.1 - 2 - 3 183 64
AK164160.1 - 2 - 3 156 64
BC039272.1 - 2 - 3 191 64
CD578615.1 - 4 - 5 479 64

View Full Product Details