Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021441.2 | NP_067416.2 | 2 - 3 | 240 | 84 |
GenBank mRNA | AK030522.1 | - | 2 - 3 | 207 | 84 |
AK052250.1 | - | 2 - 3 | 207 | 84 | |
AK087534.1 | - | 2 - 3 | 230 | 84 | |
AK154846.1 | - | 2 - 3 | 224 | 84 | |
AK156718.1 | - | 2 - 3 | 207 | 84 | |
BC051944.1 | - | 2 - 3 | 219 | 84 | |
BC058383.1 | - | 2 - 3 | 182 | 84 | |
Y09974.1 | - | 2 - 3 | 367 | 84 |