Assay Design



Gene Transcripts

Gene Symbol
Myot
Entrez Gene ID
58916
Gene Name
myotilin
Gene Aliases
5530402I04Rik, Ttid
Chromosome Location
Chr.18: 44334053 - 44355740 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033621.3 NP_001028793.1 5 - 6 946 98
GenBank mRNA AF230979.2 - 5 - 6 945 98
AK017447.1 - 5 - 6 947 98
BC016214.1 - 5 - 6 924 98
BC064052.1 - 5 - 6 919 98

View Full Product Details