Assay Design



Gene Transcripts

Gene Symbol
Gpa33
Entrez Gene ID
59290
Gene Name
glycoprotein A33 (transmembrane)
Gene Aliases
2010310L10Rik, 2210401D16Rik, BB116197
Chromosome Location
Chr.1: 166130460 - 166166510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021610.1 NP_067623.1 4 - 5 672 83
GenBank mRNA AF247659.1 - 4 - 5 672 83
BC008528.1 - 4 - 5 665 83

View Full Product Details