Assay Design



Gene Transcripts

Gene Symbol
Tlx1
Entrez Gene ID
21908
Gene Name
T cell leukemia, homeobox 1
Gene Aliases
Hox-11, Hox11
Chromosome Location
Chr.19: 45150079 - 45156943 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021901.3 NP_068701.1 2 - 3 983 87
GenBank mRNA BC018246.1 - 2 - 3 983 87

View Full Product Details