Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111075.4 | NP_001104545.1 | 3 - 4 | 603 | 60 |
NM_023117.4 | NP_075606.1 | 3 - 4 | 603 | 60 | |
GenBank mRNA | AK004836.1 | - | 3 - 4 | 560 | 60 |
AK043730.1 | - | 3 - 4 | 594 | 60 | |
AK153903.1 | - | 3 - 4 | 604 | 60 | |
AK157397.1 | - | 3 - 4 | 561 | 60 | |
BC048164.1 | - | 3 - 4 | 600 | 60 | |
BC057568.1 | - | 3 - 4 | 474 | 60 | |
BQ561430.1 | - | 3 - 4 | 259 | 60 | |
S93521.1 | - | 3 - 4 | 472 | 60 |