Assay Design



Gene Transcripts

Gene Symbol
Anxa9
Entrez Gene ID
71790
Gene Name
annexin A9
Gene Aliases
1110003P15Rik, 2310069F17Rik
Chromosome Location
Chr.3: 95296093 - 95314891 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085383.1 NP_001078852.1 9 - 10 832 62
NM_023628.2 NP_076117.2 9 - 10 811 62
GenBank mRNA AJ401160.1 - 9 - 10 765 62
AK003395.1 - 9 - 10 832 62
AK010128.1 - 9 - 10 812 62
BC062140.1 - 9 - 10 822 62

View Full Product Details