Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042499.1 | NP_001035964.1 | 7 - 8 | 666 | 107 |
GenBank mRNA | AK011280.1 | - | 7 - 8 | 608 | 107 |
AK016099.1 | - | 7 - 8 | 675 | 107 | |
AK028195.1 | - | 7 - 8 | 660 | 107 | |
AK028269.1 | - | 7 - 8 | 653 | 107 | |
BC050194.1 | - | 7 - 8 | 647 | 107 |