Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026384.3 | NP_080660.1 | 5 - 6 | 840 | 61 |
GenBank mRNA | AF384160.1 | - | 5 - 6 | 698 | 61 |
AK002443.1 | - | 5 - 6 | 845 | 61 | |
AK016660.1 | - | 5 - 6 | 789 | 61 | |
BC043447.1 | - | 5 - 6 | 840 | 61 |